Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-MEIS2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP217263
Description
MEIS2 Polyclonal specifically detects MEIS2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MEIS2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
MEIS2 | |
Recombinant protein encompassing a sequence within the center region of human MEIS2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Species reactivity based on sequence identity: Rat (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.74mg/mL | |
Western Blot 1:500-1:3000 | |
Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
Rabbit | |
42 kDa | |
0.1 mL | |
Vision | |
4212 | |
Human, Mouse, Zebrafish | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only