Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1/MEK2 Monoclonal Antibody (E.901.7), Invitrogen™
Rabbit Monoclonal Antibody
Supplier: Thermo Scientific MA515162
Description
It is not recommended to aliquot this antibody.
MEK1/MEK2 Monoclonal antibody specifically detects MEK1/MEK2 in Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Specifications
MEK1/MEK2 | |
Monoclonal | |
Unconjugated | |
MAP2K2 | |
AA589381; CFC3; CFC4; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; Dual specificity mitogen-activated protein kinase kinase 2; EC 2.7.12.2; ERK activator kinase 1; ERK activator kinase 2; hypothetical protein; hypothetical protein LOC406728; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP kinase/Erk kinase 1; Map2k1; Map2k2; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK1; MAPKK2; MEK 1; MEK 2; Mek1; MEK-1; Mek2; MEK-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; mitogen-activated protein kinase kinase type 2; MK2; MKK1; MKK2; MP2K1; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen activated, kinase 2, p45; protein kinase, mitogen-activated, ki | |
Rabbit | |
Affinity chromatography | |
RUO | |
170851, 26395, 26396, 478347, 5604, 5605, 58960, 611939 | |
-20°C | |
Liquid |
Western Blot | |
E.901.7 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P31938, P36506, P36507, Q01986, Q02750, Q1HG70, Q63932 | |
MAP2K1, MAP2K2 | |
Synthetic peptide corresponding to the sequence of human MEK1/2 | |
100 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction