Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 Mouse, Unlabeled, Clone: 25/MEK1, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610121

 View more versions of this product

Catalog No. BDB610121


Add to cart

Description

Description

MEK1 (MapK/ERK Kinase 1) is a 45kDa member of the MEK family of dual specificity kinases. MEK is activated by a variety of cellular serine/threonine kinases including c-Raf, A-Raf, c-mos, and MEK Kinase-1. Activated MEK phosphorylates MAP kinase (ERK) at threonine and tyrosine residues. This results in activation of ERK and its signaling pathway. MEK is highly specific for ERK and various MEKs preferentially phosphorylate individual ERK isoforms. MEK1 only activates ERK1 and ERK2. This specificity may result from variations in
ERK regions that are known as the phosphorylation lip and kinase backbone. MEK's localization is cytoplasmic, but mitogenic stimulation induces a mass translocation to the nucleus. Mechanisms behind this nuclear translocation remain unknown. However, MEK contains an N-terminal nuclear export signal (NES) that mediates its rapid exodus from the nucleus and restores its unstimulated cellular distribution.

Host Species: Mouse
Clone: 25
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human MEK1 aa. 2-124
Formula Weight [Chemical]: 45kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

MEK1
25/MEK1
Unconjugated
Affinity Purified
Human MEK1 aa. 2-124
50μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.