Learn More
MilliporeSigma™ anti-MEK1, Clone: C12T, Rabbit Monoclonal,
Manufacturer:  MilliporeSigma™ 05925R
Catalog No. 05-925-RMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
MEK1
C12T
Purified Rabbit Monoclonal in 0.1M Tris-Glycine (pH 7.4) with 150mM NaCl and 0.05% NaN3 with 40% glycerol.
Purified
Recombinant peptide corresponding to amino acids surrounding the TEY motif in the activation loop of human phospho-MEK1.
Protein A purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal
Human, Murine, Rat
Flow Cytometry, Western Blot
Unconjugated
NM_008927
Rabbit
IgG
100μL
Primary
EC 2.7.12.2, PRKMK1, MKK1, MAPKK1, MEK1