Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527693

Catalog No. PIPA527693


Add to cart

Description

Description

PA5-27693 targets MEK1 in IF and WB applications and shows reactivity with Human and mouse samples. The PA5-27693 immunogen is recombit fragment corresponding to a region within amino acids 1 and 215 of Human MEK1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development.
Specifications

Specifications

MEK1
Polyclonal
Unconjugated
MAP2K1
Liquid
MAP2K1
Recombit fragment corresponding to a region within amino acids 1 and 215 of Human MEK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q02750, P31938
MAPKK1, MEK1, MKK1, PRKMK1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK 1, MEK 1, dual specificity mitogen-activated protein kinase kinase 1, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26395, 5604
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit