Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA515161
Description
This antibody is predicted to react with chicken, canine, hamster, mouse, rat, rabbit and Xenopus based on sequence homology.
MAP2K1 is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development.Specifications
MEK1 | |
Polyclonal | |
MAP2K1 | |
Dual specificity mitogen-activated protein kinase kinase 1, MAP kinase kinase 1, MAPKK 1, ERK activator kinase 1, MAPK/ERK kinase 1, MEK1; Dual specificity mitogen-activated protein kinase kinase 2, MAP kinase kinase 2, MEK2, MKK2, PRKMK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5604 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q02750 | |
MAP2K1 | |
KLH conjugated synthetic peptide between 196-225 amino acids from human MEK1(MAP2K1) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction