Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Mouse, Unlabeled, Clone: 96/MEK2, BD

Mouse Monoclonal Antibody

$211.00 - $421.00

Specifications

Antigen MEK2
Isotype IgG2a
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610236
View Documents Promotion Details
BD Biosciences
610236
150μg
Each for $421.00
Add to cart
 
BDB610235
View Documents Promotion Details
BD Biosciences
610235
50μg
Each for $211.00
Add to cart
 
Description

Description

MEK (Map/Erk Kinase) 1 and 2 are serine/threonine kinases, also known as MAP kinase kinases (MAP2K1 and 2, MAPKK1 and 2, or MKK1 and 2). They activate the MAP (ctivated Protein) kinases, also known as ERKs (Extracellular signal Regulated Kinases), which are critical kinases in multiple signal transduction pathways that regulate cell growth and differentiation. Activation of MEK 1 and 2 is dependent upon phosphorylation of serines 218 and/or 222 by activated MAP kinase kinase kinases (MAP3Ks), such as the Raf isoforms. Hormones, growth and differentiating factors, or tumor promoters induce Raf activation via activation of Ras proteins. MEK2 is seven amino acids larger and shares 81% identity with MEK1. In cultured cells, MEK2 is activated by serum. In vitro, v-Raf phosphorylates and activatesMEK2. It is thought that all of these activated protein kinases are downstream of the Ras signal transduction pathway and represent an integral part of the Ras mitogenic signal.

The 96/MEK2 monoclonal antibody recognizes MEK2, regardless of phosphorylation status.

Specifications

Specifications

MEK2
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
MAP Kinase Kinase 2; ERK Kinase
IgG2a
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
96/MEK2
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit