Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141411

Catalog No. 01-675-540


Add to cart

Description

Description

This antibody will cross-react with mouse and rat MEK2, based on sequence homology. Suggested positive control: 293 whole cell lysate.

MEK2 (MAP kinase kinase/ERK kinase 2) is activated through phosphorylation of its serine residues at positions 217 and 221 by a variety of cytokines and growth factors. It is responsible for the phosphorylation/activation of MAP kinases and ERKs, and is an essential component in the transduction of mitogenic signals.
Specifications

Specifications

MEK2
Polyclonal
Unconjugated
MAP2K2
Liquid
MAP2K2
Human MEK2.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
P36507
MAPK,ERK kinase 2
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5605
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit