Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528327

Catalog No. PIPA528327


Add to cart

Description

Description

PA5-28327 targets MEK6 in WB applications and shows reactivity with Human samples. The PA5-28327 immunogen is recombit fragment corresponding to a region within amino acids 1 and 268 of Human MEK6. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein phosphorylates and activates p38 MAP kinase in response to inflammatory cytokines or environmental stress. As an essential component of p38 MAP kinase mediated signal transduction pathway, this gene is involved in many cellular processes such as stress induced cell cycle arrest, transcription activation and apoptosis.
Specifications

Specifications

MEK6
Polyclonal
Unconjugated
MAP2K6
Liquid
MAP2K6
Recombit fragment corresponding to a region within amino acids 1 and 268 of Human MEK6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P52564
MAPKK6, MEK6, MKK6, PRKMK6, SAPKK-3, SAPKK3, MAPK/ERK kinase 6, MAPKK 6, MEK 6, SAPK kinase 3, dual specificity mitogen-activated protein kinase kinase 6, protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), stress-activated protein kinase kinase 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5608
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit