Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEKK3 Mouse, Unlabeled, Clone: 40, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611103

 View more versions of this product

Catalog No. BDB611103


Add to cart

Description

Description

The Ras signaling pathway links signals from growth factor receptors to activation of the MEK/ERK kinases. This cascade of phosphorylation leads to cell growth and differentiation. External stimuli such as endotoxins, UV irradiation, heat, and hyperosmolarity, induce an array of cellular responses that culminate with gene expression. These processes are controlled by the MKK3/p38MAP kinase cascade. A third pathway responsive to environmental stress, cell death, or apoptosis is regulated by SEK/SAPKs. When overexpresssed, MEKK3 is a kinase that is capable of activating both the ERK and SAPK cascades. MEKK3 is 626 amino acids long and is ubiquitously expressed. It is positioned upstream of SEK and MEK in the signaling pathways and directly phosphorylates these enzymes. Overexpression of MEKK3 results in the activation of NFκB. Thus, MEKK3 represents an emerging family of kinases that is capable of inducing more than one signaling pathway. This indicates that specificity in the signaling cascades is probably achieved by effector kinases such as the ERKs and SAPKs.

Immunofluorescence, Western Blotting

Specifications

Specifications

MEKK3
40
Unconjugated
Affinity Purified
Human MEKK3 aa. 27-135
150μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.