Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MC4R Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific OPA115011
Description
OPA1-15011 detects MC4R from human samples. OPA1-15011 has been successfully used in immunohistochemistry procedures. By immunohistochemistry, OPA1-15011 detects MC4R in formalin fixed, paraffin embedded human tissues. The OPA1-15011 immunogen is a synthetic peptide (KLH conjugated) corresponding to the extracellular N-terminus. .
Melanocortin 4 receptor (MC4R), a Melanocortin Receptor, has been shown to affect food intake and energy balance. Recently MC4R has been shown to modulate erectile function and sexual behavior (Van Der Ploeg et al. 2002). Expression of MC4R has been reported in brain. Unlike the other melanocortin receptors, MC4R is not expressed in adrenal cortex or in melanocytes.Specifications
MC4R | |
Polyclonal | |
Unconjugated | |
P32245 | |
MC4R | |
Synthetic peptide (KLH conjugated) to the extracellular N-terminus. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
MC4R | |
MC4R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title