Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Melatonin R1A/MT1/MTNR1A, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128912

Catalog No. NBP128912


Add to cart

Description

Description

Melatonin R1A/MT1/MTNR1A Polyclonal antibody specifically detects Melatonin R1A/MT1/MTNR1A in Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Melatonin R1A/MT1/MTNR1A
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000
P48039
Mel1a receptor, MEL-1A-R, melatonin receptor 1A, melatonin receptor type 1A, MT1
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected cross-reactivity from sequence similarity: Mouse, Dog, Cow.
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
MTNR1A
Peptide with sequence C-SLKYDKLYSSKNS corresponding to internal region according to NP_005949.1.
Immunogen affinity purified
RUO
Primary
4543
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only