Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Melatonin Receptor 1B, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP136972

Catalog No. NBP136972

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Melatonin Receptor 1B
0.5 mg/mL
Western Blot 1:100 - 1:2000, Immunohistochemistry 3 - 6 ug/ml, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:32000
NP_005950.1
FGQTL2, Mel 1b melatonin receptor, Mel1b receptor, MEL-1B-R, melatonin receptor 1B, melatonin receptor MEL1B, melatonin receptor type 1B, MT2
Goat
IgG
0.1 mg
GPCR, Neuroscience
Primary
4544
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
MTNR1B
Peptide with sequence C-QDASKGSHAEGLQSP corresponding to C-Terminus according to NP_005950.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only