Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR2 Mouse anti-Human, Clone: 319701, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524041

Catalog No. PIMA524041


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit human LAIR1, recombit mouse (rm) LAIR1 or rmLAIR2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

LAIR2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q6ISS4
CD306, CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3904
Flow Cytometry, Western Blot
319701
Unconjugated
LAIR2
Lyophilized
LAIR2
Mouse myeloma cell line NS0-derived recombit human LAIR2 isoform 1 Gln22-Pro152
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit