Learn More
Mena Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610693
Description
Abl and Disabled (Dab) are essential for correct axonal connections during Drosophila development. Mutations in Enabled (Ena) rescue the genetic defects caused by mutated Abl and Dab. Mena (Mammalian Ena) is a protein with significant homology to Eva, VASP, and WASP proteins. The murine Mena gene predicts a protein of 541 amino acids. However, two additional exons that could introduce 246 amino acids onto the C-terminal region have been identified. Homology searches identified two EVH domains (Ena-Vasp homology), one near each terminus of Mena. Antibodies to Mena detect 80 kDa, 88 kDa, and 140 kDa protein with the 80 kDa and 88 kDa proteins being widely expressed and the 140 kDa protein reportedly mainly found in brain tissue. Like VASP, Mena is localized to focal contacts and, when ectopically expressed, induces the formation of F-actin outgrowths in fibroblasts.
Specifications
Mena | |
Monoclonal | |
250μg/mL | |
Mammalian Ena (Enabled) | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
21 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Chicken, Human, Mouse, Rat | |
IgA |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.