Learn More
Mena Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
$505.00
Specifications
Antigen | Mena |
---|---|
Clone | 21 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610693
|
BD Biosciences
610693 |
150 μg |
Each for $505.00
|
|
|||||
BDB610692
|
BD Biosciences
610692 |
50 μg | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
Abl and Disabled (Dab) are essential for correct axonal connections during Drosophila development. Mutations in Enabled (Ena) rescue the genetic defects caused by mutated Abl and Dab. Mena (Mammalian Ena) is a protein with significant homology to Eva, VASP, and WASP proteins. The murine Mena gene predicts a protein of 541 amino acids. However, two additional exons that could introduce 246 amino acids onto the C-terminal region have been identified. Homology searches identified two EVH domains (Ena-Vasp homology), one near each terminus of Mena. Antibodies to Mena detect 80 kDa, 88 kDa, and 140 kDa protein with the 80 kDa and 88 kDa proteins being widely expressed and the 140 kDa protein reportedly mainly found in brain tissue. Like VASP, Mena is localized to focal contacts and, when ectopically expressed, induces the formation of F-actin outgrowths in fibroblasts.
Specifications
Mena | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgA | |
Affinity Purified | |
Mammalian Ena (Enabled) |
21 | |
Western Blot | |
Unconjugated | |
RUO | |
Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.