Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Menin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121762

Catalog No. PIPA121762

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Multiple Endocrine Neoplasia is an autosomal dominant disorder characterized by a high frequency of peptic ulcer disease and primary endocrine abnormalities involving the pituitary, parathyroid, and pancreas.

PA1-21762 detects Menin in Human and Mouse samples.PA1-21762 has been successfully used in Immunoprecipitation and Western Blot procedures.PA1-21762 immunogen corresponds to Synthetic peptide (Human) - which represents a portion of the C-terminus of human MEN1..
Specifications

Specifications

Menin
Polyclonal
Unconjugated
MEN1
Purified
Rabbit
IgG
50μg
Store at 4°C
Primary
4221
Immunoprecipitation, Western Blot
1mg/mL
tris citrate, tris phosphate with 0.1% sodium azide; pH 7-8
O00255, O88559, Q9WVR8
MEN1
Synthetic peptide (Human) - which represents a portion of the C-terminus of human MEN1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit