Learn More
MOX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124468
Description
PA1-24468 detects Homeobox protein MOX-1 (MEOX1) in Human samples. PA1-24468 has been successfully used in Western Blot procedures. PA1-24468 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human MEOX1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.Specifications
MOX1 | |
Polyclonal | |
Unconjugated | |
P50221 | |
MEOX1 | |
Synthetic peptide region of human MEOX1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
MEOX1 | |
MOX1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.