Learn More
anti-Mer, Alexa Fluor 488, Clone: 125518, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8912G
Catalog No. FAB8912G
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
Mer
125518
Flow Cytometry 5 uL/10^6 cells
AAB60430
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Mouse
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Mer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Axl, rhDtk, or recombinant mouse Mer is observed.
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
MERTK
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Met1-Ala499 Accession # AAB60430
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10461
Human