Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mer, Alexa Fluor 700, Clone: 125518, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB8912N

Catalog No. FAB8912N


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: 125518
Isotype: IgG1
Species Reactivity: Hu
Immunogen: S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer, Met1-Ala499

Applications:

Flow Cytometry

Specifications

Specifications

Mer
125518
Flow Cytometry 5 uL/10^6 cells
AAB60430
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Mouse
IgG2b
100 Tests
Primary
10461
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
MERTK
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Met1-Ala499 Accession # AAB60430
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Mer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Axl, rhDtk, or recombinant mouse Mer is observed.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit