Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Mer APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB5912A
This item is not returnable.
View return policy
Description
Mer Monoclonal specifically detects Mer in Mouse samples. It is validated for Flow Cytometry.Specifications
Mer | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10461 | |
Mouse | |
Purified |
Flow Cytometry | |
108928 | |
Flow Cytometry 10 uL/10^6 cells | |
Q60805 | |
MERTK | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Mer Glu23-Phe498 Accession # Q60805 | |
100 Tests | |
Primary | |
Detects mouse Mer in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction