Learn More
anti-Mer, PE, Clone: 125518, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8912P
Catalog No. FAB8912P
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Mer
125518
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
AAB60430
c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer
Mouse
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Mer in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Axl, rhDtk, or recombinant mouse Mer is observed.
Flow Cytometry, Immunoassay
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
MERTK
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Met1-Ala499 Accession # AAB60430
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10461
Human