Learn More
Met-Enkephalin Rabbit anti-Feline, Porcine, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136172
Catalog No. 01-672-121
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
Met-enkephalin is a neurotransmitter that both competes with and mimics the effects of opiate drugs. Enkephalins play a role in a number of physiologic funtions, including pain perception and responses to stress.
Specifications
Specifications
Met-Enkephalin
Polyclonal
whole serum with 0.1% sodium azide
Methionine enkephalin; Tyr-Gly-Gly-Phe-Met-OH; [Met]enkephalin; Enkephalin M; C27H35N5O7S
Synthetic met-enkephalin conjugated to BSA.
50 μL
4° C
Primary
Feline, Porcine
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Lyophilized
Rabbit
IgG
RUO
Antibody
Polyclonal