Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Met (extracellular) Mouse anti-Human, Clone: 4F8.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051049
Description
Detect Met (extracellular) using this Anti-Met (extracellular) Antibody, clone 4F8.2 validated for use in immunohistochemistry (paraffin) and western blotting.Specifications
Met (extracellular) | |
Monoclonal | |
Unconjugated | |
NP_001120972.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunohistochemistry, Western Blot | |
4F8.2 | |
Purified mouse monoclonal in 0.1M Tris-Glycine (pH7.4), 150mM NaCl with 0.05% NaN3. | |
AUTS9, EC 2.7.10.1, HGFR , OTTHUMP00000069168, RCCP2, c-Met | |
GST-fusion protein corresponding to amino acids 80-434 of human Met. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only