Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metablastin Mouse, Unlabeled, Clone: 53, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611146

Catalog No. BDB611146


Add to cart

Description

Description

The regulation of microtubule (MT) assembly is vital to cellular processes such as organelle transport, organization of the cytoplasm, and intracellular movement of cell surface receptors. MTs are composed of tubulin subunits that exist in dynamic equilibrium between free tubulin dimers and MTs. The instability of MTs is determined by the rates of growth and shrinkage of tubulin polymers and by frequencies of transitions from growth to shrinkage (catastrophes) or from shrinkage to growth (rescues). The most well known MT regulators are the
microtubule associated proteins (MAPs) which directly bind and stabilize MTs. Metablastin (stathmin) opposes MAP activity by inducing catastrophes.Metablastin is variably phosphorylated on multiple Ser residues by kinases that are regulated by the cell cycle or by external signals. Phosphorylation of metablastin inhibits its ability to destabilize MTs and, in turn, induces tubulin polymerization. Metablastin activity is turned off during the cell cycle to allow spindle formation and cell division. Thus, metablastin is thought to function to regulate the
dynamics of MT formation in response to external signals during the interphase of the cell cycle.

Host Species: Mouse
Clone: 53
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human Metablastin aa. 38-147
Formula Weight [Chemical]: 19kDa

Bioimaging, Immunofluorescence, Western Blotting

Specifications

Specifications

Metablastin
53
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Stathmin, Metablastin
Affinity Purified
Human Metablastin aa. 38-147
50μg
Cell Biology
Primary
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit