Learn More
MilliporeSigma™ anti-Metabotropic Glutamate Receptor 2/3 ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB1553/DEL
Description
Electron Microscopy, Immunohistochemistry, Western Blotting
Specifications
Metabotropic Glutamate Receptor 2/3 | |
Unconjugated | |
Carboxy terminus peptide of rat mGluR2 conjugated to BSA with glutaraldehyde (NGREVVDSTTSSL). | |
50 μg | |
Neuroscience | |
Monkey, Rat |
Polyclonal | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary |