Learn More
Methamphetamine Sheep anti-Chemical, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA175270
Catalog No. PIPA175270
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Methamphetamine is an amphetamine with central nervous system (CNS) stimulating activity. Methamphetamine acts by both facilitating the release of catecholamines, particularly noradrenaline, dopamine and serotonin, from nerve terminals in the brain and by inhibiting their uptake. This leads to an increase in synaptic concentration of these neurotransmitters and results in increased stimulation of postsynaptic receptors. [National Center for Biotechnology Information. PubChem Compound Database; CID=10836]
Specifications
Specifications
Methamphetamine
Polyclonal
PBS with 0.09% sodium azide
Metamfetamine; D-Deoxyephedrine; D-Desoxyephedrine; D-N-Methylamphetamine; Methylamphetamine; C10H15N
Methamphetamine(p-NH2)-BSA
Purified
RUO
Antibody
Polyclonal
ELISA
Unconjugated
Liquid
Sheep
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical