Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514993

Catalog No. PIPA514993

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Methionine-tRNA ligase (EC 6.1.1.10) catalyzes the chemical reaction: ATP + L-methionine + tRNAMet <--> AMP + diphosphate + L-methionyl-tRNAMetThe 3 substrates of this enzyme are ATP, L-methionine, and tRNA(Met), whereas its 3 products are AMP, diphosphate, and L-methionyl-tRNA(Met).This enzyme participates in 3 metabolic pathways: methionine metabolism, selenoamino acid metabolism, and aminoacyl-trna biosynthesis.
Specifications

Specifications

MARS2
Polyclonal
PBS with 0.09% sodium azide
Q96GW9, Q499X9
Methionyl-tRNA synthetase, mitochondrial [Precursor]; EC 6.1.1.10; Mitochondrial methionine--tRNA ligase; MtMetRS; Methionine--tRNA ligase 2; MARS2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
212679, 92935
Immunohistochemistry, Western Blot
Unconjugated
MARS2
Liquid
MARS2
Synthetic peptide from the C-terminal region of MARS2 conjugated to KLH.
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit