Learn More
MilliporeSigma™ anti-methyl-PP2A, Clone: 2A10, ™ Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ 041479/DEL
Catalog No. 50-171-269
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Type 2 protein serine/threonine phosphatase (PP2A) is a trimer composed of a catalytic C subunit and two regulatory (A and B) subunits; many isoforms for the A and B subunits have been found. PP2A has a high specific activity against the subunit of phosphorylase kinase and is insensitive to Inhibitor-2. PP2A is distinguishable from PP2B/Calcineurin and PP2C by its partial activity in the absence of divalent cations, and by its high sensitivity to inhibition by okadaic acid. It is also thought to be the principal down-modulator of serine/threonine kinase cascades. PP2B requires Ca2+/calmodulin for activity, and is an activator of NFAT transcription factors. PP2C requires Mg2+, but is insensitive to okadaic acid, microcystin LR, or the calmodulin inhibitor trifluoperazine.
Western Blotting
Specifications
Specifications
methyl-PP2A
Unconjugated
Murine
150μg
Cell Signaling
Monoclonal
1mg/mL
Semi-purified
IgG1 κ
RUO
Primary
Chicken, Drosophila, Human, Murine, Porcine, Rabbit, Rat, Yeast