Learn More
Meteorin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130614
Catalog No. NBP130614
Add to cart
Description
Description
Meteorin Polyclonal antibody specifically detects Meteorin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
Meteorin
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 10-50 ug/ml
Purified
METRN
A synthetic peptide from the internal region of human Meteorin (METRN) conjugated to an immunogenic carrier protein was used as the immunogen.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for Meteorin.
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Sterilized water with No Preservative
C16orf23, c380A1.2, chromosome 16 open reading frame 23, meteorin, meteorin, glial cell differentiation regulator, MGC2601
Rabbit
IgG
0.5 mg
Neuroscience
Primary
79006
Reconstitute with 0.5 ml sterilized water.