Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526884

Catalog No. PIPA526884

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.
Specifications

Specifications

METTL1
Polyclonal
PBS with 0.09% sodium azide
Q9UBP6
C12orf1, TRM8, YDL201w, D1075-like gene product, methyltransferase-like 1, methyltransferase-like protein 1, tRNA (guanine(46)-N(7))-methyltransferase, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4234
Western Blot
Unconjugated
METTL1
Liquid
METTL1
KLH conjugated synthetic peptide between 147-175 amino acids from the central region of human METTL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit