Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL7A, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511518

Catalog No. PIPA511518

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

METTL7A is a methyltransferase-like protein and members of this family are SAM dependent methyltransferases.

Specifications

Specifications

METTL7A
Polyclonal
PBS with 0.09% sodium azide
Q9H8H3
Protein AAM-B
Rabbit
IgG
400μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25840
Immunohistochemistry, Western Blot
Unconjugated
METTL7A
Liquid
METTL7A
KLH conjugated synthetic peptide between 47-76 amino acids from the N-terminal region of human METTL7A
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit