Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

microfibrillar-associated protein 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human MFAP3 protein.

Manufacturer:  Abnova Corporation H00004238B01

Catalog No. 89-005-754


Add to cart

Description

Description

Sequence: MKLHCCLFTLVASIIVPAAFVLEDVDFDQMVSLEANRSSYNASFPSSFELSASSHSDDDVIIAKEGTSVSIECLLTASHYEDVHWHNSKGQQLDGRSRGGKWLVSDNFLNITNVAFDDRGLYTCFVTSPIRASYSVTLRVIFTSGDMSVYYMIVCLIAFTITLILNVTRLCMMSSHLRKTEKAINEFFRTEGAEKLQKAFEIAKRIPIITSAKTLELAKVTQFKTMEFARYIEELARSVPLPPLILNCRAFVEEMFEAVRVDDPDDLGERIKERPALNAQGGIYVINPEMGRSNSPGGDSDDGSLNEQGQEIAVQVSVHLQSETKSIDTESQGSSHFSPPDDIGSAESNCNYKDGAYENCQL
Specifications

Specifications

microfibrillar-associated protein 3
Unconjugated
No additive
NM_005927.3
DKFZp586F0219
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4238
Western Blot
Mouse polyclonal antibody raised against a full-length human MFAP3 protein.
MFAP3
Serum
MFAP3
MFAP3 (NP_005918.1, 1 a.a. ~ 362 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit