Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFN2, Mouse, Clone: 4F5, Abnova™

Mouse monoclonal antibody raised against a partial recombinant MFN2.

Manufacturer:  Abnova Corporation H00009927M07

Catalog No. 89-002-766


Add to cart

Description

Description

This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified. [provided by RefSeq

Sequence: FKRQFVEHASEKLQLVISYTGSNCSHQVQQELSGTFAHLCQQVDVTRENLEQEIAAMNKKIEVLDSLQSKAKLLRNKAGWLDSELNMFTHQYLQPSR
Specifications

Specifications

MFN2,
4F5
Mouse monoclonal antibody raised against a partial recombinant MFN2.
MFN2
Liquid
MFN2
MFN2 (NP_055689, 661 a.a. ~ 757 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Western Blot
Unconjugated
In 1X PBS, pH 7.4
NM_014874
CMT2A|CMT2A2|CPRP1|HSG|KIAA0214|MARF
Mouse
IgG2a Kappa
RUO
Primary
9927
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit