Learn More
SLC25A28 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525168
Catalog No. PIPA525168
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MFRN2 is mitochondrial iron transporter that mediates iron uptake. It is probably required for heme synthesis of hemoproteins and Fe-S cluster assembly in non-erythroid cells. The iron delivered into the mitochondria, presumably as Fe(2+), is then probably delivered to ferrochelatase to catalyze Fe(2+) incorporation into protoprophyrin IX to make heme.
Specifications
Specifications
SLC25A28
Polyclonal
Unconjugated
SLC25A28
Liquid
SLC25A28
KLH conjugated synthetic peptide between 36-64 amino acids from the N-terminal region of human MFRN2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide
Q96A46
MFRN2, MRS3/4, MRS4L, hMRS3/4, mitochondrial RNA splicing protein 3/4, mitochondrial RNA-splicing protein 3/4 homolog, mitochondrial iron transporter 2, mitoferrin-2, putative mitochondrial solute carrier
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
81894