Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526927

Catalog No. PIPA526927


Add to cart

Description

Description

This antibody is predicted to react with mouse and porcine based on sequence homology.

Specifications

Specifications

MFSD6
Polyclonal
PBS with 0.09% sodium azide
Q6ZSS7
MMR2, hMMR2, macrophage MHC class I receptor 2 homolog, macrophage MHC receptor 2, major facilitator superfamily domain-containing protein 6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54842
Western Blot
Unconjugated
MFSD6
Liquid
MFSD6
KLH conjugated synthetic peptide between 656-685 amino acids from the C-terminal region of human MFSD6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit