Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dehydrogenase/reductase (SDR family) member 11 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human MGC4172 protein.
Manufacturer: Abnova Corporation H00079154B01
Description
Sequence: MFSAIRSQHSGVDICINNAGLARPDTLLSGSTSGWKDMFNVNVLALSICTREAYQSMKERNVDDGHIININSMSGHRVLPLSVTHFYSATKYAVTALTEGLRQELREAQTHIRATCISPGVVETQFAFKLHDKDPEKAAATYEQMKCLKPEDVAEAVIYVLSTPAHIQIGDIQMRPTEQVTSpecifications
dehydrogenase/reductase (SDR family) member 11 | |
Unconjugated | |
No additive | |
BC002731 | |
ARPG836|FLJ39232|MGC4172|SDR24C1 | |
Mouse | |
50 uL | |
Stem Cell Biology | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human MGC4172 protein. | |
DHRS11 | |
Serum | |
DHRS11 | |
MGC4172 (AAH02731, 1 a.a. ~ 181 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
79154 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title