Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
magnesium-dependent phosphatase 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MGC5987 protein.
Manufacturer: Abnova Corporation H00145553B01
Description
Sequence: MARLPKLAVFDLDYTLWPFWVDTHVDPPFHKSSDGTVRDRRGQDVRLYPEVPEVLKRLQSLGVPGAAASRTSEIEGANQLLELFDLFRYFVHREIYPGSKITHFERLQQKTGIPFSQMIFFDDERRNIVDVSKLGVTCIHIQNGMNLQTLSQGLETFAKAQTGPLRSSLEESPFEASpecifications
magnesium-dependent phosphatase 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MGC5987 protein. | |
MDP-1 | |
MGC5987 | |
Mouse | |
50 μL | |
Yes | |
145553 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
NM_138476.2 | |
MDP-1 | |
MGC5987 (NP_612485.2, 1 a.a. ∼ 176 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title