Learn More
mGluR6/mGluR7, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA121039
Catalog No. PIPA121039
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.
PA1-21039 detects Metabotropic Glutamate Receptor 6 & 7 in Human, Mouse, Rat and Non-human primate samples.PA1-21039 has been successfully used in Immunohistochemistry (Paraffin), Immunoprecipitation and Western blot procedures. For immunohistochemistry (paraffin), staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 minutes.PA1-21039 immunogen corresponds to synthetic peptide (commercially sensitive amino acid sequence) (Human) (C-terminal).
Specifications
Specifications
mGluR6 + mGluR7
Polyclonal
Unconjugated
GRM7
Purified
GRM6, GRM7
Synthetic peptide (commercially sensitive amino acid sequence) (Human) (C-terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Non-human Primate, Rat
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
O15303, Q14831, Q5NCH9, Q68ED2, P35349, P35400
Grm6, Grm7, Metabotropic glutamate receptor 6, Metabotropic glutamate receptor 7, mGluR6, mGluR7
Rabbit
IgG
500μL
4° C, do not freeze
Primary
2916, 2917