Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR6/mGluR7, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121040

Catalog No. PIPA121040

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: Ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR17, NMDA13, KAI2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.

PA1-21040 detects Metabotropic Glutamate Receptor 6 & 7 in Human, Mouse, Rat and Non-human primate samples.PA1-21040 has been successfully used in Immunohistochemistry (Paraffin) procedures.PA1-21040 immunogen corresponds to synthetic peptide (Human) (C-terminal).
Specifications

Specifications

mGluR6 + mGluR7
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
O15303, Q14831, Q5NCH9, Q68ED2, P35349, P35400
Grm6, Grm7, Metabotropic glutamate receptor 6, Metabotropic glutamate receptor 7, mGluR6, mGluR7
Rabbit
IgG
7mL
4° C, do not freeze
Primary
2916, 2917
Immunohistochemistry (Paraffin)
Unconjugated
GRM7
Purified
GRM6, GRM7
Synthetic peptide (Human) (C terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Non-human Primate, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit