Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR6 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132350

Catalog No. PIPA132350


Add to cart

Description

Description

PA1-32350 detects mGluR6 from rat samples. PA1-32350 has been successfully used in Western blot applications. The PA1-32350 immunogen is: C-terminus of rat mGluR6. AAPPQNENAEDAK Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

has adenylate cyclase inhibiting metabotropic glutamate receptor activity; may play a role in synaptic transmission between photoreceptor cells and ON-bipolar cells in the retina.
Specifications

Specifications

mGluR6
Polyclonal
Unconjugated
Grm6
Liquid
GRM6
C-terminus of rat mGluR6. AAPPQNENAEDAK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
tris glycine with 0.1% BSA and 0.1% sodium azide
P35349
mGluR6
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
24419
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit