Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-G Mouse anti-Human, APC, Clone: MEM-G/9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA119465

Catalog No. 01-676-200


Add to cart

Description

Description

This antibody reacts with native form of human HLA-G1 on the cell surface as well as with soluble HLA-G5 isoform in its beta2-microglobulin associated form. Reactivity with HLA-G3 was also reported. It will not cross-react with mouse.

HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail.
Specifications

Specifications

HLA-G
Monoclonal
0.1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide
P17693
DADB-15K14.8, MHC-G
Mouse
IgG1
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
3135
Flow Cytometry
MEM-G/9
APC
HLA-G
Liquid
HLA-G
Recombit human HLA-G refolded with beta- 2 Microglobulin and peptide.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit