Learn More
Mi-2b (CHD4) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061306
Catalog No. 06-130-6MI
Add to cart
Description
Description
Use Anti-Mi-2b (CHD4) Antibody (rabbit polyclonal antibody) validated in western blotting to detect Mi-2b (CHD4) also known as ATP-dependent helicase CHD4, Mi-2 autoantigen 218 kDa protein, chromodomain helicase DNA binding protein 4.
Specifications
Specifications
Mi-2b (CHD4)
Unconjugated
Serum
Rabbit
Unpurified
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Polyclonal
Human, Murine
Western Blot
Unpurified rabbit polyclonal in serum with 0.05% sodium azide.
CHD4; CHD-4; Mi-2b; Mi2-BETA; Mi2-beta
GST-tagged recombinant protein corresponding to mouse CHD-4 at and around the N-terminus.
100μL
Primary
NP_666091.1