Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIC-1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514186
Description
Colon cancer-associated protein Mic1.Specifications
MIC-1 | |
Polyclonal | |
C18orf8 | |
Uncharacterized protein C18orf8; Colon cancer-associated protein Mic1; Mic-1; C18orf8 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
29919 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q96DM3 | |
C18ORF8 | |
KLH conjugated synthetic peptide between 399-428 amino acids from the central region of human MIC1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction