Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MICAL2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526602

Catalog No. PIPA526602


Add to cart

Specifications

Specifications

MICAL2
Polyclonal
Unconjugated
MICAL2
Liquid
MICAL2
KLH conjugated synthetic peptide between 55-83 amino acids from the N-terminal region of human MICAL2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Western Blot
2.0 mg/mL
PBS with 0.09% sodium azide
O94851
MICAL2PV1, MICAL2PV2, flavoprotein oxidoreductase MICAL2, protein MICAL-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9645
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit