Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICB APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems FAB1599A025
This item is not returnable.
View return policy
Description
MICB Monoclonal specifically detects MICB in Human samples. It is validated for Flow Cytometry.Specifications
MICB | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4277 | |
Human | |
Purified |
Flow Cytometry | |
236511 | |
Flow Cytometry 10 uL/10^6 cells | |
CAI18747 | |
MICB | |
Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 | |
25 Tests | |
Primary | |
Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction