Learn More
anti-MICB, PE, Clone: 236511, R&D Systems™
Manufacturer:  R&D Systems FAB1599P025
 View more versions of this product
Catalog No. FAB1599P025
Add to cart
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
MICB
236511
Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells
CAI18747
MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog
Mouse
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human MICB in direct ELISAs and Western blots. Does not cross-react with recombinant human MICA.
Flow Cytometry, Immunoassay
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
MICB
Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4277
Human