Learn More
MilliporeSigma™ anti-Microtubule-Associated Protein 2 (MAP2); Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB5622/DEL
Catalog No. 50-173-875
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
The microtubule-associated protein 2 (MAP2) is an abundant neuronal cytoskeletal protein that binds to tubulin and stabilizes microtubules. (Herzog and Weber, 1978). MAP2 is essential for the development and maitenance of neuronal morphology (Matus, 1991). In neurons MAP2 occurs as three primary isoforms, the high molecular weight MAP2a, MAP2b, and the low molecular weight MAP2c, that result from alternative splicing of the MAP2 gene (Chung et al., 1996). The low molecular weight isoform, MAP2c, is expressed in developing brain and is down-regulated during brain maturation, whereas the high molecular weight MAP2b is expressed in both developing and adult brain. The MAP2a appears only after brain maturation (Tucker, 1990). All these forms bind to microtubules through a domain near the carboxyl terminus that contains either three or four similar repeats of a 31-amino-acid motif (Lewis et al., 1988). MAP2 together with MAP4 and tau proteins belong to the family of thermostable proteins associated with microtubules.
ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Microtubule-Associated Protein 2 (MAP2)
Semi-purified
Purified Microtubule-associated protein from rat brain.
RUO
Primary
Human, Murine, Rat
Unconjugated
Rabbit
100μL
Neuroscience
Polyclonal