Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523166

Catalog No. PIPA523166


Add to cart

Alert:

d


Description

Description

MAP1 LC3A is a structural protein and a component of both MAP1A and MAP1B microtubule-binding complexes which mediate the physical interactions between microtubules and components of the cytoskeleton. A member of the MAP1LC3 family, LC3A is known to have an ubiquitin fold at the C-terminal region and 2 helices at the N-terminal region. The posttranslational modification of LC3A precursor molecule leads to cleavage by APG4B/ATG4B, resulting in the cytosolic form, LC3-I which is further activated and modified to form the membrane-bound form, LC3-II. LC3A, a mammalian homolog of yeast Atg8, plays an essential role in autophagy, leading to bulk degradation of cytoplasmic components by the lysosomal system where it acts as an adaptor protein between microtubules and autophagosomes. Abundant expression of LC3A is seen in heart, brain, liver, skeletal muscle and testis but absent in thymus and peripheral blood leukocytes.
Specifications

Specifications

LC3A
Polyclonal
Unconjugated
MAP1LC3A
Liquid
MAP1LC3A
Synthetic peptide (1 PSDRPFKQRRSFAD 15) of Microtubule-associated proteins 1A/1B light chain 3A precursor (MAP1 LC3A) protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat, Zebrafish
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q2HJ23, Q9H492, Q91VR7, Q6XVN8
RP11-346K17.1, ATG8E, LC3, LC3A, MAP1ALC3, MAP1BLC3, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, autophagy-related protein LC3 A, autophagy-related ubiquitin-like modifier LC3 A, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
362245, 406466, 514547, 66734, 84557
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit