Learn More
anti-Midkine, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131158
Catalog No. NBP131158
Add to cart
Description
Description
Midkine Polyclonal antibody specifically detects Midkine in Human samples. It is validated for Western Blot.
Specifications
Specifications
Midkine
0.33mg/mL
Western Blot 1:100-1:1000
Affinity Purified
MDK
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Midkine. The exact sequence is proprietary.
16 kDa
0.1 ml
Cell Cycle and Replication
Primary
4192
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.